accessibility menu, dialog, popup

UserName

H3COOHOOH3COCH3

CAS RN 33130-04-0

IUPAC Name: 3-(2,3,4-trimethoxyphenyl)propanoic acid
Synonyms: 3-(2,3,4-trimethoxyphenyl)propanoic acid
Molecular Weight (g/mol): 240.26
Molecular Formula: C12H16O5
InChi Key: QOPNYPCVRBRZOP-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCC(O)=O)C(OC)=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results