accessibility menu, dialog, popup

UserName

CAS RN 331-64-6

IUPAC Name: 2-fluoro-4-methoxybenzaldehyde
Synonyms: 2-fluoro-4-methoxybenzaldehyde
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: UNWQNFJBBWXFBG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results