accessibility menu, dialog, popup

UserName

HO(R)(S)HO(R)OOOHOH(R)OHHOO(E)

CAS RN 327-97-9

IUPAC Name: (1S,3R,4R,5R)-3-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}-1,4,5-trihydroxycyclohexane-1-carboxylic acid
Synonyms: chlorogenic acid acid, chlorogenic 3-trans-caffeoylquinic acid hlorogenate acid, 3-caffeoylquinic heriguard hlorogenic acid 3-O-caffeoylquinic acid 3 caffeoylquinic acid chlorogenic acid (8CI)
Molecular Weight (g/mol): 354.31
Molecular Formula: C16H18O9
InChi Key: CWVRJTMFETXNAD-JUHZACGLSA-N
SMILES: O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O
Quantity 
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)