accessibility menu, dialog, popup

UserName

ONOFFNOO

CAS RN 327-92-4

IUPAC Name: 1,5-difluoro-2,4-dinitrobenzene
Synonyms: 1,5-difluoro-2,4-dinitrobenzene
Molecular Weight (g/mol): 204.09
Molecular Formula: C6H2F2N2O4
InChi Key: VILFTWLXLYIEMV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=C(F)C=C1F)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results