accessibility menu, dialog, popup

UserName

CAS RN 32604-29-8

CH3OOOCH3O

CAS RN 32604-29-8

IUPAC Name: 4'-(acetyloxy)-[1,1'-biphenyl]-4-yl acetate
Synonyms: 4'-(acetyloxy)-[1,1'-biphenyl]-4-yl acetate
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: RQMBBMQDXFZFCC-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C(C=C1)C1=CC=C(OC(C)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results