accessibility menu, dialog, popup

UserName

FFFOOS

CAS RN 326-91-0

IUPAC Name: 4,4,4-trifluoro-1-(thiophen-2-yl)butane-1,3-dione
Synonyms: thenoyltrifluoroacetone ttfa cpd TTA TTB thenoyltrifluoromethylacetone 2-thenoyltrifluoroacetone TTFA α-thenoyltrifluoroacetone
Molecular Weight (g/mol): 222.18
Molecular Formula: C8H5F3O2S
InChi Key: TXBBUSUXYMIVOS-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)CC(=O)C1=CC=CS1
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)