CAS RN 326-06-7
IUPAC Name:
4,4,4-trifluoro-1-phenylbutane-1,3-dione
Synonyms:
4,4,4-trifluoro-1-phenylbutane-1,3-dione
Molecular Weight (g/mol):
216.16
Molecular Formula:
C10H7F3O2
InChi Key:
VVXLFFIFNVKFBD-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(=O)CC(=O)C1=CC=CC=C1
1
–
2
of
2
results
1
3-Benzoyl-1,1,1-trifluoroacetone, 98+%
CAS: 326-06-7 Molecular Formula: C10H7F3O2 Molecular Weight (g/mol): 216.16 MDL Number: MFCD00000425 InChI Key: VVXLFFIFNVKFBD-UHFFFAOYSA-N Synonym: benzoyl-1,1,1-trifluoroacetone,benzoyltrifluoroacetone,4,4,4-trifluoro-1-phenyl-1,3-butanedione,benzoyl trifluoroacetyl methane,3-benzoyl-1,1,1-trifluoroacetone,1,3-butanedione, 4,4,4-trifluoro-1-phenyl,4,4,4-trifluoro-1-phenyl-butane-1,3-dione,1-benzoyl-3,3,3-trifluoroacetone,.omega.-trifluoroacetyl acetophenone,1-benzoyl-3,3,3-trifluoro-2-propanone PubChem CID: 67589 IUPAC Name: 4,4,4-trifluoro-1-phenylbutane-1,3-dione SMILES: FC(F)(F)C(=O)CC(=O)C1=CC=CC=C1