CAS RN 31972-52-8
CAS RN 31972-52-8
IUPAC Name:
2-(2-{[(tert-butoxy)carbonyl]amino}acetamido)acetic acid
Synonyms:
{2-[(tert-butoxycarbonyl)amino]acetamido}acetic acid
Molecular Weight (g/mol):
232.24
Molecular Formula:
C9H16N2O5
InChi Key:
HWBAHOVOSOAFLE-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCC(=O)NCC(O)=O
1
–
1
of
1
results
1
N-Boc-glycylglycine, 97%, Thermo Scientific Chemicals
CAS: 31972-52-8 Molecular Formula: C9H16N2O5 Molecular Weight (g/mol): 232.236 MDL Number: MFCD00038558 InChI Key: HWBAHOVOSOAFLE-UHFFFAOYSA-N Synonym: boc-gly-gly-oh,boc-gly-gly,n-tert-butoxycarbonyl glycylglycine,n-2-n-boc-amino-acetyl-glycine,boc-glycylglycine,2-2-tert-butoxycarbonyl amino acetamido acetic acid,2-tert-butoxycarbonyl amino acetamido acetic acid,2-2-tert-butoxycarbonylamino acetamido acetic acid,2-2-tert-butoxy carbonyl amino acetamido acetic acid,t-boc-glygly PubChem CID: 333466 IUPAC Name: 2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]acetic acid SMILES: CC(C)(C)OC(=O)NCC(=O)NCC(=O)O