accessibility menu, dialog, popup

UserName

CAS RN 3164-85-0

KH3CCH3OO

CAS RN 3164-85-0

IUPAC Name: potassium 2-ethylhexanoate
Synonyms: potassium 2-ethylhexanoate
Molecular Weight (g/mol): 182.3
Molecular Formula: C8H15KO2
InChi Key: ZUFQCVZBBNZMKD-UHFFFAOYNA-M
SMILES: [K+].CCCCC(CC)C([O-])=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)