CAS RN 31545-32-1
CAS RN 31545-32-1
IUPAC Name:
(3,5-dichloropyridin-2-yl)thiourea
Synonyms:
3,5-dichloropyridin-2-ylthiourea
Molecular Weight (g/mol):
222.09
Molecular Formula:
C6H5Cl2N3S
InChi Key:
RRWWUBPSZSTRRC-UHFFFAOYSA-N
SMILES:
NC(=S)NC1=C(Cl)C=C(Cl)C=N1
1
–
1
of
1
results
1
N-(3,5-Dichloro-2-pyridyl)thiourea, 97%, Thermo Scientific™
CAS: 31545-32-1 Molecular Formula: C6H5Cl2N3S Molecular Weight (g/mol): 222.087 MDL Number: MFCD15525457 InChI Key: RRWWUBPSZSTRRC-UHFFFAOYSA-N Synonym: 3,5-dichloro-pyridin-2-yl-thiourea,n-3,5-dichloro-2-pyridyl thiourea PubChem CID: 18506847 IUPAC Name: (3,5-dichloropyridin-2-yl)thiourea SMILES: C1=C(C=NC(=C1Cl)NC(=S)N)Cl