accessibility menu, dialog, popup

UserName

CAS RN 315-22-0

CH3(R)OO(R)NOO(R)CH3OH(R)CH3OH(R)

CAS RN 315-22-0

IUPAC Name: (1R,4R,5R,6R,16R)-5,6-dihydroxy-4,5,6-trimethyl-2,8-dioxa-13-azatricyclo[8.5.1.0¹³,¹⁶]hexadec-10-ene-3,7-dione
Synonyms: monocrotaline crotaline monocrotalin testosterone oenanthate (all-xi)-monocrotaline (all-xi)-isomer monocrotaline monocrotaline (substance) monocrotaline, (all-xi)-isomer monocrotaline, (all-xi)- ccris 416
Molecular Weight (g/mol): 325.36
Molecular Formula: C16H23NO6
InChi Key: QVCMHGGNRFRMAD-XFGHUUIASA-N
SMILES: C[C@H]1C(=O)O[C@@H]2CCN3CC=C(COC(=O)[C@](C)(O)[C@]1(C)O)[C@H]23
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results