accessibility menu, dialog, popup

UserName

CAS RN 31489-18-6

NNOO

CAS RN 31489-18-6

IUPAC Name: 4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzonitrile
Synonyms: 4-(2,5-dioxopyrrol-1-yl)benzonitrile
Molecular Weight (g/mol): 198.18
Molecular Formula: C11H6N2O2
InChi Key: PRXNTCCKSPNEFB-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)