accessibility menu, dialog, popup

UserName

NBrNHSH2N

CAS RN 31430-38-3

IUPAC Name: (5-bromopyridin-2-yl)thiourea
Synonyms: 5-bromopyridin-2-ylthiourea
Molecular Weight (g/mol): 232.1
Molecular Formula: C6H6BrN3S
InChi Key: BWEBPIDYSNIWEB-UHFFFAOYSA-N
SMILES: NC(=S)NC1=NC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results