accessibility menu, dialog, popup

UserName

CAS RN 3102-56-5

H3COHNH2OH

CAS RN 3102-56-5

IUPAC Name: 2-aminooctadecane-1,3-diol
Synonyms: 2-aminooctadecane-1,3-diol
Molecular Weight (g/mol): 301.52
Molecular Formula: C18H39NO2
InChi Key: OTKJDMGTUTTYMP-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCC(O)C(N)CO
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results