CAS RN 30897-90-6
IUPAC Name:
magnesium(2+) 4-fluoro-2-methylbenzen-1-ide bromide
Synonyms:
magnesium(2+) 4-fluoro-2-methylbenzen-1-ide bromide
Molecular Weight (g/mol):
213.33
Molecular Formula:
C7H6BrFMg
InChi Key:
OCGINFOPBMDLBD-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].CC1=CC(F)=CC=[C-]1