accessibility menu, dialog, popup

UserName

CH3NH2

CAS RN 3082-64-2

IUPAC Name: 1-phenylpropan-1-amine
Synonyms: 1-phenylpropan-1-amine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: AQFLVLHRZFLDDV-UHFFFAOYNA-N
SMILES: CCC(N)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results