CAS RN 30766-03-1
CAS RN 30766-03-1
IUPAC Name:
4-bromopyridine-2-carboxylate
Synonyms:
4-bromopyridine-2-carboxylate
Molecular Weight (g/mol):
201
Molecular Formula:
C6H3BrNO2
InChi Key:
RPHHYRNGCJYQSP-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC(Br)=CC=N1
1
–
1
of
1
results
1
4-Bromopyridine-2-carboxylic acid, 97%
CAS: 30766-03-1 Molecular Formula: C6H4BrNO2 Molecular Weight (g/mol): 202.007 MDL Number: MFCD03426665 InChI Key: RPHHYRNGCJYQSP-UHFFFAOYSA-N Synonym: 4-bromopicolinic acid,4-bromo-2-pyridinecarboxylic acid,4-bromo-pyridinecarboxylic acid,2-pyridinecarboxylic acid, 4-bromo,4-bromo-pyridine-2-carboxylic acid,aurora 23242,pubchem4064,acmc-209hio,ksc222g5r PubChem CID: 735150 IUPAC Name: 4-bromopyridine-2-carboxylic acid SMILES: C1=CN=C(C=C1Br)C(=O)O