CAS RN 30742-62-2
IUPAC Name:
2-(morpholin-4-yl)-5-nitrobenzaldehyde
Synonyms:
2-(morpholin-4-yl)-5-nitrobenzaldehyde
Molecular Weight (g/mol):
236.23
Molecular Formula:
C11H12N2O4
InChi Key:
PCGDWIWUQDHQLK-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(C=O)=C(C=C1)N1CCOCC1