accessibility menu, dialog, popup

UserName

CAS RN 306937-21-3

IUPAC Name: 5-(5-chloro-1,2,4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride
Synonyms: 5-(5-chloro-1,2,4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 301.17
Molecular Formula: C6H2Cl2N2O2S3
InChi Key: DCLCBWFFYZNVES-UHFFFAOYSA-N
SMILES: ClC1=NC(=NS1)C1=CC=C(S1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results