accessibility menu, dialog, popup

UserName

NHNOFFF

CAS RN 306936-65-2

IUPAC Name: 5-[4-(trifluoromethyl)phenyl]-1H-pyrazole-4-carbaldehyde
Synonyms: 3-[4-(trifluoromethyl)phenyl]-2H-pyrazole-4-carbaldehyde
Molecular Weight (g/mol): 240.19
Molecular Formula: C11H7F3N2O
InChi Key: PMKPJAJMBQBTKE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C1=C(C=O)C=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results