accessibility menu, dialog, popup

UserName

OOOFFF

CAS RN 306936-00-5

IUPAC Name: 5-[2-(trifluoromethoxy)phenyl]furan-2-carbaldehyde
Synonyms: 5-[2-(trifluoromethoxy)phenyl]furan-2-carbaldehyde
Molecular Weight (g/mol): 256.18
Molecular Formula: C12H7F3O3
InChi Key: UTGWIIGCXFJJHZ-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1C1=CC=C(O1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results