accessibility menu, dialog, popup

UserName

CAS RN 306935-19-3

CH3NSONOHNH2(E/Z)

CAS RN 306935-19-3

IUPAC Name: N'-hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzene-1-carboximidamide
Synonyms: N'-hydroxy-4-[(2-methyl-1,3-thiazol-4-yl)methoxy]benzenecarboximidamide
Molecular Weight (g/mol): 263.32
Molecular Formula: C12H13N3O2S
InChi Key: QHRPKGPTUYZTBY-UHFFFAOYSA-N
SMILES: CC1=NC(COC2=CC=C(C=C2)C(N)=NO)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results