accessibility menu, dialog, popup

UserName

NSSH2N

CAS RN 306934-91-8

IUPAC Name: 5-(pyridin-2-yl)thiophene-2-carbothioamide
Synonyms: 5-(pyridin-2-yl)thiophene-2-carbothioamide
Molecular Weight (g/mol): 220.31
Molecular Formula: C10H8N2S2
InChi Key: VOYHCOULKKDLRP-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C(S1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results