CAS RN 3059-97-0
IUPAC Name:
(2R)-2-azaniumyl-2-methylbutanoate
Synonyms:
(2R)-2-ammonio-2-methylbutanoate
Molecular Weight (g/mol):
117.15
Molecular Formula:
C5H11NO2
InChi Key:
GCHPUFAZSONQIV-RXMQYKEDSA-N
SMILES:
CC[C@@](C)([NH3+])C([O-])=O