accessibility menu, dialog, popup

UserName

H3COOONOOCH3ClH2N

CAS RN 305834-79-1

IUPAC Name: ethyl 2-amino-6-chloro-4-(1-cyano-2-ethoxy-2-oxoethyl)-4H-chromene-3-carboxylate
Synonyms: ethyl 2-amino-6-chloro-4-(1-cyano-2-ethoxy-2-oxoethyl)-4H-chromene-3-carboxylate
Molecular Weight (g/mol): 364.78
Molecular Formula: C17H17ClN2O5
InChi Key: DXVKFBGVVRSOLI-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C#N)C1C2=C(OC(N)=C1C(=O)OCC)C=CC(Cl)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results