CAS RN 304448-55-3
IUPAC Name:
3-hydroxy-N'-[(3-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]naphthalene-2-carbohydrazide
Synonyms:
3-hydroxy-N'-[(3-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]naphthalene-2-carbohydrazide
Molecular Weight (g/mol):
322.32
Molecular Formula:
C18H14N2O4
InChi Key:
NRQJUIHYFNUXFT-UHFFFAOYSA-N
SMILES:
OC1=CC(=CNNC(=O)C2=CC3=CC=CC=C3C=C2O)C=CC1=O
1
–
2
of
2
results
1
R&D Systems™ Dynasore
CAS: 304448-55-3 Molecular Formula: C18H14N2O4
2
Dynamin Inhibitor I, Dynasore, TRC
CAS: 304448-55-3 Molecular Formula: C18 H14 N2 O4 Molecular Weight (g/mol): 322.315 Synonym: 3-Hydroxy-[(3,4-dihydroxyphenyl)methylene]hydrazide 2-Naphthalenecarboxylic Acid IUPAC Name: N-[(3,4-dihydroxyphenyl)methylideneamino]-3-hydroxynaphthalene-2-carboxamide SMILES: Oc1ccc(C=NNC(=O)c2cc3ccccc3cc2O)cc1O