CAS RN 302803-72-1
IUPAC Name:
3-(2,4-dichlorophenyl)-3-oxo-2-(4-oxo-1,2,3,4-tetrahydroquinazolin-2-ylidene)propanenitrile
Synonyms:
3-(2,4-dichlorophenyl)-3-oxo-2-(4-oxo-1,3-dihydroquinazolin-2-ylidene)propanenitrile
Molecular Weight (g/mol):
358.18
Molecular Formula:
C17H9Cl2N3O2
InChi Key:
SESYPWCSIZUIAS-UHFFFAOYSA-N
SMILES:
ClC1=CC(Cl)=C(C=C1)C(=O)C(C#N)=C1NC(=O)C2=CC=CC=C2N1
1
–
1
of
1
results
1
R&D Systems™ Ciliobrevin A
A Tocris product, part of R&D Systems. Hedgehog (Hh) pathway antagonist, inhibits ciliogenesis; also inhibits dynein