CAS RN 30269-04-6
IUPAC Name:
pyrene-1,8-diamine
Synonyms:
pyrene-1,8-diamine
Molecular Weight (g/mol):
232.29
Molecular Formula:
C16H12N2
InChi Key:
BLYOXQBERINFDU-UHFFFAOYSA-N
SMILES:
NC1=CC=C2C=CC3=CC=C(N)C4=CC=C1C2=C34
1
–
2
of
2
results
1
1,8-Pyrenediamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,8-Pyrenediamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.