accessibility menu, dialog, popup

UserName

H3COOCH3(Z)(E)

CAS RN 3025-30-7

IUPAC Name: ethyl (2E,4Z)-deca-2,4-dienoate
Synonyms: ethyl (2E,4Z)-2,4-decadienoate ethyl(E,Z)-decadienoate ethyl decadienoate
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: OPCRGEVPIBLWAY-QNRZBPGKSA-N
SMILES: CCCCC\C=C/C=C/C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results