CAS RN 30186-18-6
IUPAC Name:
1-(4-bromo-2-hydroxyphenyl)ethan-1-one
Synonyms:
acetophenone,2-hydroxy-4-bromo
Molecular Weight (g/mol):
215.05
Molecular Formula:
C8H7BrO2
InChi Key:
LQCMMXGKEGWUIM-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(O)C=C(Br)C=C1
1
–
1
of
1
results
1
4'-Bromo-2'-hydroxyacetophenone, 95%
CAS: 30186-18-6 Molecular Formula: C8H7BrO2 Molecular Weight (g/mol): 215.05 MDL Number: MFCD03428533 InChI Key: LQCMMXGKEGWUIM-UHFFFAOYSA-N Synonym: 1-4-bromo-2-hydroxyphenyl ethanone,4'-bromo-2'-hydroxyacetophenone,4-bromo-2-hydroxyacetophenone,1-4-bromo-2-hydroxyphenyl ethan-1-one,acetophenone,2-hydroxy-4-bromo,ethanone, 1-4-bromo-2-hydroxyphenyl,2-acetyl-5-bromophenol,1-4-bromo-2-hydroxy-phenyl ethanone,1-4-bromo-2-hydroxy-phenyl-ethanone PubChem CID: 10727403 IUPAC Name: 1-(4-bromo-2-hydroxyphenyl)ethanone SMILES: CC(=O)C1=C(O)C=C(Br)C=C1