CAS RN 301-12-2
IUPAC Name:
dimethyl {[2-(ethanesulfinyl)ethyl]sulfanyl}phosphonate
Synonyms:
oxydemeton methyl demeton-methyl sulphoxide caswell no. 455 metilmercaptofosoksid (ussr) methyl oxydemeton (substance) demeton-S methyl sulfoxide demeton-S-methyl sulfoxide metaisosystox sulfoxide metasystox-R demeton-S-methyl-sulfoxid
Molecular Weight (g/mol):
246.28
Molecular Formula:
C6H15O4PS2
InChi Key:
PMCVMORKVPSKHZ-UHFFFAOYNA-N
SMILES:
CCS(=O)CCSP(=O)(OC)OC