CAS RN 300574-36-1
IUPAC Name:
5-bromo-4,5,6,7-tetrahydro-2,1,3-benzoxadiazol-4-one
Synonyms:
5-bromo-6,7-dihydro-5H-2,1,3-benzoxadiazol-4-one
Molecular Weight (g/mol):
217.02
Molecular Formula:
C6H5BrN2O2
InChi Key:
RLBJBTYYLDXIHR-UHFFFAOYNA-N
SMILES:
BrC1CCC2=NON=C2C1=O