CAS RN 300557-74-8
IUPAC Name:
2-chloro-1-[2,5-dimethyl-1-(4-methylphenyl)-1H-pyrrol-3-yl]ethan-1-one
Synonyms:
2-chloro-1-[2,5-dimethyl-1-(4-methylphenyl)pyrrol-3-yl]ethanone
Molecular Weight (g/mol):
261.75
Molecular Formula:
C15H16ClNO
InChi Key:
BAEXXXPZKCRCQD-UHFFFAOYSA-N
SMILES:
CC1=CC(C(=O)CCl)=C(C)N1C1=CC=C(C)C=C1