CAS RN 29682-12-0
IUPAC Name:
1-[4-(benzyloxy)-2-hydroxyphenyl]ethan-1-one
Synonyms:
1-[4-(benzyloxy)-2-hydroxyphenyl]ethanone
Molecular Weight (g/mol):
242.27
Molecular Formula:
C15H14O3
InChi Key:
AGQNLHOTLJFJCG-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1O
1
–
4
of
4
results
1
2-Hydroxy-4-benzyloxyacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-Hydroxy-4-benzyloxyacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-Hydroxy-4-benzyloxyacetophenone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.