accessibility menu, dialog, popup

UserName

CAS RN 29681-45-6

IUPAC Name: methyl 5-methylpyridine-3-carboxylate
Synonyms: methyl 5-methylpyridine-3-carboxylate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: KQILMMLAGGFMCM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=CC(C)=C1
Molecular Weight (g/mol) 
  • (9)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (6)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (9)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (6)
  • (2)