CAS RN 29557-51-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific™ High–Purity Fos-Choline Detergents
Thermo Scientific™ High–Purity Fos-Choline Detergents
Fos-choline detergents are a class of zwitterionic detergents commonly used in protein solubilization for various applications, including membrane protein research, protein purification, and structural biology studies.
2
n-Dodecylphosphocholine, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.