accessibility menu, dialog, popup

UserName

CAS RN 2949-22-6

H3COONCO

CAS RN 2949-22-6

IUPAC Name: ethyl 2-isocyanatoacetate
Synonyms: ethyl 2-isocyanatoacetate
Molecular Weight (g/mol): 129.12
Molecular Formula: C5H7NO3
InChi Key: DUVOZUPPHBRJJO-UHFFFAOYSA-N
SMILES: CCOC(=O)CN=C=O
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)