accessibility menu, dialog, popup

UserName

BrNN

CAS RN 294877-33-1

IUPAC Name: 1-(3-bromophenyl)-1H-pyrazole
Synonyms: 1-(3-bromophenyl)pyrazole
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: KUWAIOZTKMAIPI-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results