CAS RN 29156-65-8
IUPAC Name:
sodium tetrachlorogolduide hydrate
Synonyms:
sodium hydrate tetrachloroaurate
Molecular Weight (g/mol):
379.77
Molecular Formula:
AuCl4H2NaO
InChi Key:
VTMCVHXKFJBNKC-UHFFFAOYSA-J
SMILES:
O.[Na+].Cl[Au-](Cl)(Cl)Cl
1
–
1
of
1
results
1
Sodium tetrachloroaurate hydrate, 98%
CAS: 29156-65-8 Molecular Formula: AuCl4Na Molecular Weight (g/mol): 361.76 MDL Number: MFCD00149161 InChI Key: IXPWAPCEBHEFOV-UHFFFAOYSA-J Synonym: sodium tetrachloroaurate iii hydrate,aucl4.na.h2o,sodium tetrachloroaurate 1--water 1/1/1 PubChem CID: 18410391 IUPAC Name: sodium;tetrachlorogold(1-);hydrate SMILES: [Na+].Cl[Au-](Cl)(Cl)Cl