CAS RN 28797-48-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
11-(Chloroacetyl)-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one, TRC
CAS: 28797-48-0 Molecular Formula: C14H10ClN3O2 Molecular Weight (g/mol): 287.7 IUPAC Name: 11-(2-chloroacetyl)-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one SMILES: ClCC(=O)N1c2ccccc2C(=O)Nc3cccnc13