accessibility menu, dialog, popup

UserName

OONHO

CAS RN 287930-73-8

IUPAC Name: 4-benzyl-2-hydroxymorpholin-3-one
Synonyms: 4-benzyl-2-hydroxymorpholin-3-one
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: CAGSEMPCDRYWFN-UHFFFAOYNA-N
SMILES: OC1OCCN(CC2=CC=CC=C2)C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results