accessibility menu, dialog, popup

UserName

CAS RN 287917-97-9

NHNBrO

CAS RN 287917-97-9

IUPAC Name: 4-bromo-1H-pyrazole-5-carbaldehyde
Synonyms: 4-bromo-2H-pyrazole-3-carbaldehyde
Molecular Weight (g/mol): 174.99
Molecular Formula: C4H3BrN2O
InChi Key: UWGFONONBAIDAF-UHFFFAOYSA-N
SMILES: BrC1=C(NN=C1)C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results