accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNOO

CAS RN 286961-15-7

IUPAC Name: benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine-1-carboxylate
Synonyms: benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
Molecular Weight (g/mol): 343.23
Molecular Formula: C19H26BNO4
InChi Key: QDSFHRPYZPQWEJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CCN(CC1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results