CAS RN 28664-35-9
CAS RN 28664-35-9
IUPAC Name:
3-hydroxy-4,5-dimethyl-2,5-dihydrofuran-2-one
Synonyms:
sotolon
Molecular Weight (g/mol):
128.13
Molecular Formula:
C6H8O3
InChi Key:
UNYNVICDCJHOPO-UHFFFAOYNA-N
SMILES:
CC1OC(=O)C(O)=C1C
1
–
3
of
3
results
1
4,5-Dimethyl-3-hydroxy-2(5H)-furanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4,5-Dimethyl-3-hydroxy-2(5H)-furanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4,5-Dimethyl-3-hydroxy-2(5H)-furanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.