accessibility menu, dialog, popup

UserName

ClSOONHOHNO

CAS RN 28485-18-9

IUPAC Name: 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride
Synonyms: 2,4-dioxo-1,3-dihydropyrimidine-5-sulfonyl chloride
Molecular Weight (g/mol): 210.59
Molecular Formula: C4H3ClN2O4S
InChi Key: FQHNJCWWBKYCJW-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CNC(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results