accessibility menu, dialog, popup

UserName

H3CH3COOOOCH3CH3OO

CAS RN 2847-00-9

IUPAC Name: 5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-6-one
Synonyms: 5-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-dihydro-3aH-furo[2,3-d][1,3]dioxol-6-one
Molecular Weight (g/mol): 258.27
Molecular Formula: C12H18O6
InChi Key: QSRFRZJIRAOQSJ-UHFFFAOYNA-N
SMILES: CC1(C)OCC(O1)C1OC2OC(C)(C)OC2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results