accessibility menu, dialog, popup

UserName

CH3H3CCH3OONH(S)OOF

CAS RN 284493-72-7

IUPAC Name: (3S)-3-{[(tert-butoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoate
Synonyms: (3S)-3-[(tert-butoxycarbonyl)amino]-3-(4-fluorophenyl)propanoate
Molecular Weight (g/mol): 282.29
Molecular Formula: C14H17FNO4
InChi Key: WRVBNEFIXONNFA-NSHDSACASA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC([O-])=O)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results