CAS RN 28224-73-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,3:4,6-Di-O-benzylidene-D-mannitol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,3:4,6-Di-O-benzylidene-D-mannitol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.