accessibility menu, dialog, popup

UserName

CH3H3CCH3OONSH2N

CAS RN 274682-80-3

IUPAC Name: tert-butyl 3-carbamothioylpiperidine-1-carboxylate
Synonyms: tert-butyl 3-carbamothioylpiperidine-1-carboxylate
Molecular Weight (g/mol): 244.35
Molecular Formula: C11H20N2O2S
InChi Key: ZCELPXRDWJCIJE-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC(C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results