CAS RN 271-29-4
IUPAC Name:
1H-pyrrolo[2,3-c]pyridine
Synonyms:
6-azaindole
Molecular Weight (g/mol):
118.14
Molecular Formula:
C7H6N2
InChi Key:
XLKDJOPOOHHZAN-UHFFFAOYSA-N
SMILES:
N1C=CC2=CC=NC=C12
1
–
1
of
1
results
1
6-Azaindole, 97+%
CAS: 271-29-4 Molecular Formula: C7H6N2 Molecular Weight (g/mol): 118.14 MDL Number: MFCD01686899 InChI Key: XLKDJOPOOHHZAN-UHFFFAOYSA-N Synonym: 6-azaindole,1h-pyrrolo 2,3-c pyridine,harmyrine,1,6-diazaindene,6h-pyrrolo 2,3-c pyridine,6-azaindol,pyrrolo 2,3-c pyridine,6-aza-6h-indole,zlchem 439 PubChem CID: 9219 IUPAC Name: 1H-pyrrolo[2,3-c]pyridine SMILES: N1C=CC2=CC=NC=C12